Invitrogen
Manufacturer Code:PA5106788
Catalog # PIPA5106788
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TEAD3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DTEF-1; DTEF-1 ETFR-1 ETF-related factor 1 Tcf13r2 TEA domain family member 3 TEA domain family member 5 TEA domain transcription factor 3 Tead3 TEAD-3 TEAD5 TEF5 TEF-5 transcriptional enhancer factor 5 Transcriptional enhancer factor TEF-5 transcriptional enhancer factor TEF-5 (DTEF-1); ETF-related factor 1; ETFR-1; TEA domain family member 3; TEA domain family member 5; TEAD-3; transcriptional enhancer factor 5; Transcriptional enhancer factor TEF-5; transcriptional enhancer factor TEF-5 (DTEF-1)
Gene Aliases: DTEF-1; ETFR-1; Tcf13r2; TEAD-3; TEAD3; TEAD5; TEF-5; TEF5
UniProt ID: (Human) Q99594, (Mouse) P70210
Entrez Gene ID: (Human) 7005, (Mouse) 21678
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.