Novus Biologicals
Manufacturer Code:NBP183949
Catalog # NBP183949
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:QDIKPFAQPAYPIQPPLPPTLSSYEPLAPLPSAAASVPVWQDRTIASSRLRLLEYSAFMEVQRDPDTYSKHLFVHIGQTNPAFSDPPLEAVDVRQIYDKFPEK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DTEF-1; DTEF-1 ETFR-1 TEA domain family member 3TEAD-3 TEAD5TEF5TEA domain family member 5 TEF-5 transcriptional enhancer factor 5 transcriptional enhancer factor TEF-5 Transcriptional enhancer factor TEF-5 (DTEF-1); TEA domain family member 3; TEA domain family member 5; TEAD-3; transcriptional enhancer factor 5; Transcriptional enhancer factor TEF-5; transcriptional enhancer factor TEF-5 (DTEF-1)
Gene Aliases: DTEF-1; ETFR-1; TEAD-3; TEAD3; TEAD5; TEF-5; TEF5
UniProt ID: (Human) Q99594
Entrez Gene ID: (Human) 7005
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.