Novus Biologicals
Manufacturer Code:NBP29312820UL
Catalog # NB028145
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 130-330 of human TEAD3 (NP_003205.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DTEF-1; DTEF-1 ETFR-1 TEA domain family member 3TEAD-3 TEAD5TEF5TEA domain family member 5 TEF-5 transcriptional enhancer factor 5 transcriptional enhancer factor TEF-5 Transcriptional enhancer factor TEF-5 (DTEF-1); TEA domain family member 3; TEA domain family member 5; TEAD-3; transcriptional enhancer factor 5; Transcriptional enhancer factor TEF-5; transcriptional enhancer factor TEF-5 (DTEF-1)
Gene Aliases: DTEF-1; ETFR-1; TEAD-3; TEAD3; TEAD5; TEF-5; TEF5
UniProt ID: (Human) Q99594
Entrez Gene ID: (Human) 7005
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.