Proteintech
Manufacturer Code:211591AP150UL
Catalog # 50-100-2786
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TEAD2 Fusion Protein Ag14074 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Embryonic TEA domain-containing factor; ETEF-1; ETF; ETF TEA domain family member 2 TEAD 2 TEAD2 TEF 4 TEF4; TEA domain family member 2; TEAD-2; Transcriptional enhancer factor TEF-4
Gene Aliases: Etdf; ETF; TEAD-2; TEAD2; TEF-4; TEF4
UniProt ID: (Human) Q15562, (Mouse) P48301
Entrez Gene ID: (Human) 8463, (Mouse) 21677
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.