Proteintech
Manufacturer Code:252391AP150UL
Catalog # 50-173-4807
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TDRD5 Fusion Protein Ag18896 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RP11 427G13.1 TDRD5 tudor domain containing 5 TUDOR3; testicular tissue protein Li 194; tudor domain containing protein 5; Tudor domain-containing protein 5
Gene Aliases: Gm103; TDRD5; TUDOR3
UniProt ID: (Human) Q8NAT2, (Mouse) Q5VCS6
Entrez Gene ID: (Human) 163589, (Mouse) 214575
Molecular Function: nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.