Novus Biologicals
Manufacturer Code:NBP274487G
Catalog # NBP274487G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4H1 |
Immunogen | Full length recombinant protein of human TDP1 (NP_060789) produced in HEK293T cells. |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: EC 3.1.4 EC 3.1.4.- FLJ11090 MGC104252 SCAN1 Tyr-DNA phosphodiesterase 1 tyrosyl-DNA phosphodiesterase 1; Tyr-DNA phosphodiesterase 1; Tyrosyl-DNA phosphodiesterase 1
Gene Aliases: TDP1
UniProt ID: (Human) Q9NUW8
Entrez Gene ID: (Human) 55775
Molecular Function:
hydrolase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.