Invitrogen
Manufacturer Code:PA598189
Catalog # PIPA598189
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human Tyrosyl-DNA phosphodiesterase 1 protein (1-260AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: protein expressed in male leptotene and zygotene spermatocytes 501; Tyr-DNA phosphodiesterase 1; tyrosyl-DNA phodphodiesterase 1; Tyrosyl-DNA phosphodiesterase 1
Gene Aliases: 2810481F14Rik; 4921509N21Rik; AI838772; AW493413; E430034L06Rik; MLZ-501; SCAN1; TDP1
UniProt ID: (Human) Q9NUW8, (Rat) Q4G056
Entrez Gene ID: (Human) 55775, (Rat) 314380, (Mouse) 104884
Molecular Function: hydrolase phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.