Invitrogen
Manufacturer Code:PA572009
Catalog # PIPA572009
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 212-239 amino acids from the Central region of human TDP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: protein expressed in male leptotene and zygotene spermatocytes 501; Tyr-DNA phosphodiesterase 1; tyrosyl-DNA phodphodiesterase 1; Tyrosyl-DNA phosphodiesterase 1; Tyrosyl-DNA phosphodiesterase 1 Tyr-DNA phosphodiesterase 1 314- TDP1
Gene Aliases: 2810481F14Rik; 4921509N21Rik; AI838772; AW493413; E430034L06Rik; MLZ-501; SCAN1; TDP1
UniProt ID: (Human) Q9NUW8
Entrez Gene ID: (Human) 55775, (Mouse) 104884
Molecular Function:
hydrolase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.