Invitrogen
Manufacturer Code:PA5116017
Catalog # PIPA5116017
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TDP1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2810481F14Rik 4921509N21Rik AI838772 AW493413 E430034L06Rik MLZ-501 protein expressed in male leptotene and zygotene spermatocytes 501 SCAN1 Tdp1 tyr-DNA phosphodiesterase 1 tyrosyl-DNA phodphodiesterase 1 tyrosyl-DNA phosphodiesterase 1; protein expressed in male leptotene and zygotene spermatocytes 501; Tyr-DNA phosphodiesterase 1; tyrosyl-DNA phodphodiesterase 1; Tyrosyl-DNA phosphodiesterase 1
Gene Aliases: 2810481F14Rik; 4921509N21Rik; AI838772; AW493413; E430034L06Rik; MLZ-501; SCAN1; TDP1
UniProt ID: (Human) Q9NUW8, (Rat) Q4G056
Entrez Gene ID: (Human) 55775, (Rat) 314380, (Mouse) 104884
Molecular Function: hydrolase phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.