Proteintech
Manufacturer Code:128921AP150UL
Catalog # 50-554-983
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TDP-43 (C-terminal) Fusion Protein Ag4003 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ALS10 TAR DNA binding protein TAR DNA binding protein 43 TARDBP TDP 43 TDP43; TAR DNA-binding protein 43; TAR DNA-binding protein-43; TDP-43
Gene Aliases: 1190002A23Rik; ALS10; C85084; TARDBP; TDP-43; TDP43
UniProt ID: (Human) Q13148, (Mouse) Q921F2
Entrez Gene ID: (Human) 23435, (Mouse) 230908, (Rat) 298648
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.