Invitrogen
Manufacturer Code:MA531571
Catalog # PIMA531571
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GT6310 |
Immunogen | Recombit protein encompassing a sequence within the center region of human TDP43. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ALS10 antibody TAR DNA binding protein antibody TARDBP antibody TDP-43 antibody TDP43 antibody; TAR DNA-binding protein 43; TAR DNA-binding protein-43; TDP-43
Gene Aliases: 1190002A23Rik; ALS10; C85084; TARDBP; TDP-43; TDP43
UniProt ID: (Human) Q13148, (Mouse) Q921F2
Entrez Gene ID: (Human) 23435, (Mouse) 230908, (Rat) 298648
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.