Invitrogen
Manufacturer Code:711051
Catalog # PI711051
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Oligoclonal |
Type | Antibody |
Clone | 1HCLC |
Immunogen | Protein corresponding to Human TARDBP (aa 130-275) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage,avoiding freeze/thaw cycles.,store at -20°C |
Rabbit Recombinant Polyclonal Antibody
For Research Use Only
Protein Aliases: TAR DNA-binding protein 43; TAR DNA-binding protein-43; TDP-43
Gene Aliases: ALS10; TARDBP; TDP-43; TDP43
UniProt ID: (Human) Q13148
Entrez Gene ID: (Human) 23435
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.