Invitrogen
Manufacturer Code:PA577018
Catalog # PIPA577018
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human TDO2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: TDO; tdo variant1; tdo variant2; TDO2; TO; TRPO; Tryptamin 2,3-dioxygenase; Tryptophan 2,3-dioxygenase; Tryptophan 23-dioxygenase TDO Tryptamin 23-dioxygenase Tryptophan oxygenase TO TRPO Tryptophan pyrrolase Tryptophanase TDO2 TDO; Tryptophan oxygenase; Tryptophan pyrrolase; tryptophan-2,3-dioxygenase; Tryptophanase
Gene Aliases: AA407491; chky; TDO; TDO2; TO; TPH2; TRPO
UniProt ID: (Human) P48775, (Mouse) P48776
Entrez Gene ID: (Human) 6999, (Mouse) 56720
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.