Proteintech
Manufacturer Code:158801AP150UL
Catalog # 50-173-0549
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TDO2 Fusion Protein Ag8642 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: TDO; TDO TDO2 TO TPH2 TRPO Tryptamin 23 dioxygenase tryptophan 23 dioxygenase Tryptophan oxygenase Tryptophan pyrrolase Tryptophanase; tdo variant1; tdo variant2; TDO {ECO:0000255|HAMAP-Rule:MF_03020}; TDO2; TO; TO {ECO:0000255|HAMAP-Rule:MF_03020}; TRPO; TRPO {ECO:0000255|HAMAP-Rule:MF_03020}; Tryptamin 2,3-dioxygenase; tryptamin 2,3-dioxygenase {ECO:0000255|HAMAP-Rule:MF_03020}; Tryptophan 2,3-dioxygenase; tryptophan 2,3-dioxygenase A; tryptophan 2,3-dioxygenase {ECO:0000255|HAMAP-Rule:MF_03020}; Tryptophan oxygenase; tryptophan oxygenase {ECO:0000255|HAMAP-Rule:MF_03020}; Tryptophan pyrrolase; tryptophan pyrrolase {ECO:0000255|HAMAP-Rule:MF_03020}; tryptophan-2,3-dioxygenase; tryptophan-23-dioxygenase; Tryptophanase; tryptophanase {ECO:0000255|HAMAP-Rule:MF_03020}
Gene Aliases: AA407491; chky; TDO; TDO2; TO; TPH2; TRPO
UniProt ID: (Human) P48775, (Mouse) P48776, (Rat) P21643
Entrez Gene ID: (Human) 6999, (Mouse) 56720, (Rat) 64206
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.