Novus Biologicals
Manufacturer Code:NBP243595
Catalog # NBP243595
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1142 |
Immunogen | Recombinant protein encompassing a sequence within the center region of human TDG. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 3.2.2.29 thymine-DNA glycosylaseG/T mismatch-specific thymine DNA glycosylase; G/T mismatch-specific thymine DNA glycosylase; hTDG; Thymine-DNA glycosylase
Gene Aliases: hTDG; TDG
UniProt ID: (Human) Q13569
Entrez Gene ID: (Human) 6996
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.