Proteintech
Manufacturer Code:133701AP150UL
Catalog # 50-172-8686
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TDG Fusion Protein Ag4190 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-JUN leucine zipper interactive protein JZA-3; G/T mismatch-specific thymine DNA glycosylase; hTDG; TDG thymine DNA glycosylase; Thymine-DNA glycosylase
Gene Aliases: E130317C12Rik; hTDG; JZA-3; Jza1; TDG
UniProt ID: (Human) Q13569, (Mouse) Q3TPW4
Entrez Gene ID: (Human) 6996, (Rat) 114521, (Mouse) 21665
Molecular Function: DNA binding protein DNA glycosylase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.