Invitrogen
Manufacturer Code:PA599729
Catalog # PIPA599729
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TDE1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA959945 AA960047 Aigp1 AIGP-1 axotomy-induced glyco/Golgi protein 1 Axotomy-induced glycoprotein 1 Diff33 Membrane protein TMS-1 SBBI99 SERINC3 serine incorporator 3 TDE Tde1 Tms1 TMS-1 tumor differentially expressed 1 tumor differentially expressed protein 1; Serine incorporator 3; Tumor differentially expressed protein 1
Gene Aliases: AIGP1; DIFF33; SBBI99; SERINC3; TDE; TDE1; TMS-1
UniProt ID: (Human) Q13530
Entrez Gene ID: (Human) 10955
Molecular Function:
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.