Invitrogen
Manufacturer Code:PA530417
Catalog # PIPA530417
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 247 and 467 of Human TDE1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AIGP1 DIFF33 SBBI99 TDE TDE1 TMS-1 placental transmembrane protein (mouse testicular tumor differentially expressed) transmembrane protein SBBI99 tumor differentially expressed 1 tumor differentially expressed protein 1 tumour differentially expressed 1; Serine incorporator 3; Tumor differentially expressed protein 1
Gene Aliases: AIGP1; DIFF33; SBBI99; SERINC3; TDE; TDE1; TMS-1
UniProt ID: (Human) Q13530
Entrez Gene ID: (Human) 10955
Molecular Function:
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.