Invitrogen
Manufacturer Code:MA534861
Catalog # PIMA534861
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JE48-48 |
Immunogen | Synthetic peptide within C-terminal human TCP1 alpha. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 65 kDa antigen AI528772 BOS_10000 c-cpn CCT Cct1 Ccta CCTalpha CCT-alpha D6S230E p63 RCJMB04_2b5 Tailless complex polypeptide 1A Tailless complex polypeptide 1B t-complex 1 T-complex protein 1 subunit alpha TCP1 Tcp-1 TCP-1-A TCP-1-alpha TCP-1-B Tp63 TRiC YD8142.13 YD8142B.04 YDR212W; CCT-alpha; T-complex protein 1 subunit alpha; T-complex protein 1, alpha subunit; tailless complex polypeptide 1; TCP-1-alpha
Gene Aliases: CCT-alpha; CCT1; CCTA; D6S230E; TCP-1-alpha; TCP1
UniProt ID: (Human) P17987
Entrez Gene ID: (Human) 6950
Molecular Function: chaperone chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.