Invitrogen
Manufacturer Code:MA191679
Catalog # PIMA191679
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 23c |
Immunogen | Purified recombit mouse TCP1 alpha construct encoding the C-terminal half of the 1.8 kb full-length Tcp 1b gene expressed in E. coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: 65 kDa antigen; AI528772 CCT Cct1 Ccta TRic Tcp-1 Tp63 c-cpn p63; CCT-alpha; chaperonin-containing T-complex polypeptide alpha subunit; cytosolic chaperonin containing t-complex polypeptide 1; t-complex polypeptide 1; T-complex protein 1 alpha subunit-like protein; T-complex protein 1 subunit alpha; T-complex protein 1 subunit alpha A; T-complex protein 1 subunit alpha B; Tailless complex polypeptide 1A; Tailless complex polypeptide 1B; TCP-1-A; TCP-1-alpha; TCP-1-B
Gene Aliases: AI528772; c-cpn; CCT; CCT-alpha; CCT1; Ccta; CCTalpha; p63; Tcp-1; TCP1; Tp63; TRiC
UniProt ID: (Bovine) Q32L40, (Rat) P28480, (Mouse) P11983, (Chinese hamster) P18279
Entrez Gene ID: (Rabbit) 100302435, (Dog) 484064, (Bovine) 512043, (Sheep) 100135437, (Rat) 24818, (Mouse) 21454, (Chinese hamster) 100689476
Molecular Function: chaperone chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.