Invitrogen
Manufacturer Code:PA518251
Catalog # PIPA518251
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (SYEDAVHSGALND) corresponding to the C-terminus amino acids of TCP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CCT-alpha; T-complex protein 1 subunit alpha; T-complex protein 1, alpha subunit; tailless complex polypeptide 1; TCP-1-alpha; TCP1 t-complex 1 CCT-alpha CCT1 CCTa D6S230E TCP-1-alpha T-complex protein 1 T-complex protein 1 alpha subunit tailless complex polypeptide 1
Gene Aliases: CCT-alpha; CCT1; CCTA; D6S230E; TCP-1-alpha; TCP1
UniProt ID: (Human) P17987
Entrez Gene ID: (Human) 6950
Molecular Function: chaperone chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.