Invitrogen
Manufacturer Code:PA5102531
Catalog # PIPA5102531
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TCP1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 65 kDa antigen AI528772 c-cpn CCT Cct1 Ccta CCTalpha CCT-alpha chaperonin-containing T-complex alpha subunit TCP1 chaperonin-containing T-complex polypeptide alpha subunit cytosolic chaperonin containing t-complex polypeptide 1 D6S230E p63 RCJMB04_2b5 tailless complex polypeptide 1 tailless complex polypeptide 1A Tailless complex polypeptide 1B t-complex 1 t-complex polypeptide 1 t-complex protein 1 T-complex protein 1 alpha subunit-like protein T-complex protein 1 subunit alpha T-complex protein 1 subunit alpha A T-complex protein 1 subunit alpha B T-complex protein 1 alpha subunit Tcp1 Tcp-1 TCP-1-A TCP-1-alpha TCP-1-B Tp63 TRic Unknown (protein for MGC:133746) YD8142.13 YD8142B.04 YDR212W; CCT-alpha; t-complex polypeptide 1; T-complex protein 1 subunit alpha; T-complex protein 1 subunit alpha A; T-complex protein 1 subunit alpha B; T-complex protein 1, alpha subunit; tailless complex polypeptide 1; Tailless complex polypeptide 1A; Tailless complex polypeptide 1B; TCP-1-A; TCP-1-alpha; TCP-1-B
Gene Aliases: AI528772; c-cpn; CCT; CCT-alpha; CCT1; CCTA; CCTalpha; D6S230E; p63; Tcp-1; TCP-1-alpha; TCP1; Tp63; TRic
UniProt ID: (Human) P17987, (Rat) P28480, (Mouse) P11983
Entrez Gene ID: (Human) 6950, (Rat) 24818, (Mouse) 21454
Molecular Function: chaperone chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.