Invitrogen
Manufacturer Code:PA528200
Catalog # PIPA528200
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 117 and 417 of Human TCP1 zeta |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acute morphine dependence related protein 2; Acute morphine dependence-related protein 2; amino acid transport defect-complementing; CCT-zeta CCT-zeta-1 CCT6 Cctz HTR3 MoDP-2 TCP-1-zeta TCP20 TCPZ TTCP20 T-complex protein 1 subunit zeta T-complex protein 1 zeta subunit acute morphine dependence related protein 2 acute morphine dependence-related protein 2 amino acid transport defect-complementing chaperonin containing T-complex subunit 6 histidine transp; CCT-zeta-1; chaperonin containing T-complex subunit 6; chaperonin containing TCP-1; chaperonin containing TCP1, subunit 6A (zeta 1); chaperonin subunit 6a (zeta); histidine transport regulator 3; HTR3; T-complex protein 1 subunit zeta; T-complex protein 1, zeta subunit; TCP-1-zeta; Tcp20
Gene Aliases: CCT-zeta; CCT-zeta-1; CCT6; CCT6A; CCTZ; Cctz-1; Cctz1; HTR3; MoDP-2; TCP-1-zeta; TCP20; TCPZ; TTCP20
UniProt ID: (Human) P40227, (Mouse) P80317
Entrez Gene ID: (Human) 908, (Mouse) 12466
Molecular Function:
chaperone
chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.