Invitrogen
Manufacturer Code:PA596289
Catalog # PIPA596289
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 248-497 of human CCT8. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCT-theta; Chaperonin containing T-complex polypeptide 1 subunit 8; chaperonin containing TCP1, subunit 8 (theta); chaperonin subunit 8 (theta); Renal carcinoma antigen NY-REN-15; T-complex protein 1 subunit theta; TCP-1-theta
Gene Aliases: AI132397; C21orf112; CCT8; CCTQ; D21S246; KIAA0002; PRED71; Tcpq
UniProt ID: (Human) P50990, (Mouse) P42932
Entrez Gene ID: (Human) 10694, (Rat) 288305, (Mouse) 12469
Molecular Function: chaperone chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.