Invitrogen
Manufacturer Code:PA569389
Catalog # PIPA569389
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of human TCP-1 theta |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bdav; bette davis; C21orf112 Cctq D21S246 PRED71 CCT-theta T-complex protein 1 subunit theta T-complex protein 1 theta subunit TCP-1-theta renal carcinoma antigen NY-REN-15; CCT-theta; Chaperonin containing T-complex polypeptide 1 subunit 8; chaperonin containing TCP1, subunit 8 (theta); chaperonin subunit 8 (theta); Renal carcinoma antigen NY-REN-15; T-complex protein 1 subunit theta; TCP-1-theta
Gene Aliases: AI132397; C21orf112; CCT8; CCTQ; D21S246; fa22h09; KIAA0002; PRED71; Tcpq; wu:fa22h09; zgc:56059
UniProt ID: (Human) P50990, (Bovine) Q3ZCI9, (Mouse) P42932
Entrez Gene ID: (Horse) 100053750, (Human) 10694, (Dog) 478399, (Rabbit) 100353291, (Bovine) 281047, (Rat) 288305, (Mouse) 12469, (Guinea pig) 100713457, (Zebrafish) 394037
Molecular Function: chaperone chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.