Invitrogen
Manufacturer Code:PA530403
Catalog # PIPA530403
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 284 and 527 of Human TCP1 theta |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C21orf112 Cctq D21S246 PRED71 CCT-theta T-complex protein 1 subunit theta T-complex protein 1 theta subunit TCP-1-theta renal carcinoma antigen NY-REN-15; CCT-theta; Chaperonin containing T-complex polypeptide 1 subunit 8; chaperonin containing TCP1, subunit 8 (theta); Renal carcinoma antigen NY-REN-15; T-complex protein 1 subunit theta; TCP-1-theta
Gene Aliases: C21orf112; CCT8; CCTQ; D21S246; KIAA0002; PRED71
UniProt ID: (Human) P50990
Entrez Gene ID: (Human) 10694
Molecular Function: chaperone chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.