Invitrogen
Manufacturer Code:MA190088
Catalog # PIMA190088
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | PK/8/4/4i/2f |
Immunogen | TCP1 beta peptide conjugated to purified tuberculin protein derivative (PPD) (Mouse). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: AI528772 CCT Cct1 Ccta TRic Tcp-1 Tp63 c-cpn p63; CCT-beta; chaperonin containing t-complex polypeptide 1, beta subunit; chaperonin containing t-complex polypeptide 1, subunit 2; chaperonin containing TCP1, subunit 2 (beta); chaperonin subunit 2 (beta); chaperonin-containing T-complex polypeptide beta subunit; epididymis secretory sperm binding protein Li 100n; T-complex protein 1 subunit beta; T-complex protein 1, beta subunit; TCP-1-beta
Gene Aliases: 99D8.1; CCT-beta; CCT2; CCTB; HEL-S-100n; PRO1633; TCP-1-beta
UniProt ID: (Bovine) Q3ZBH0, (Human) P78371, (Mouse) P80314, (Rat) Q5XIM9
Entrez Gene ID: (Bovine) 505313, (Pig) 100157776, (Rabbit) 100216334, (Dog) 474445, (Human) 10576, (Sheep) 101102222, (Mouse) 12461, (Chinese hamster) 100759253, (Rat) 299809
Molecular Function: chaperone chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.