Invitrogen
Manufacturer Code:MA535560
Catalog # PIMA535560
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1097 |
Immunogen | A synthesized peptide derived from human TCP1 beta |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 99D8.1 AI528772 BOS_5233 c-cpn CCT Cct1 Cct2 Ccta CCTB CCT-beta HEL-S-100n p63 PRO1633 T-complex protein 1 subunit beta Tcp-1 TCP-1-beta Tp63 TRic; CCT-beta; chaperonin containing t-complex polypeptide 1, beta subunit; chaperonin containing t-complex polypeptide 1, subunit 2; chaperonin containing TCP1, subunit 2 (beta); chaperonin subunit 2 (beta); epididymis secretory sperm binding protein Li 100n; T-complex protein 1 subunit beta; T-complex protein 1, beta subunit; TCP-1-beta
Gene Aliases: 99D8.1; CCT-beta; CCT2; CCTB; HEL-S-100n; PRO1633; TCP-1-beta
UniProt ID: (Human) P78371, (Mouse) P80314, (Rat) Q5XIM9
Entrez Gene ID: (Human) 10576, (Mouse) 12461, (Rat) 299809
Molecular Function: chaperone chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.