Invitrogen
Manufacturer Code:MA531253
Catalog # PIMA531253
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 49 |
Immunogen | Recombit Mouse TCN2 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: II TC TC2 TC-2 TCII TC II D22S676 D22S750; TC II; TC-2; TCII; Transcobalamin II; Transcobalamin-2
Gene Aliases: AW208754; Tcn-2; Tcn2
UniProt ID: (Mouse) O88968
Entrez Gene ID: (Mouse) 21452
Molecular Function: cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.