Proteintech
Manufacturer Code:663771IG150UL
Catalog # 50-173-6806
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2G6D2 |
Immunogen | TCN2 Fusion Protein Ag2825 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: macrocytic anemia; TC 2 TC II TC2 TCII TCN2 Transcobalamin 2 Transcobalamin II; TC II; TC-2; Transcobalamin II; transcobalamin II; macrocytic anemia; Transcobalamin-2; vitamin B12-binding protein 2
Gene Aliases: D22S676; D22S750; II; TC; TC II; TC-2; TC2; TCII; TCN2
UniProt ID: (Human) P20062
Entrez Gene ID: (Human) 6948
Molecular Function: cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.