Invitrogen
Manufacturer Code:PA5100347
Catalog # PIPA5100347
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TCN2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AW208754 D22S676 D22S750 II macrocytic anemia TC TC II TC TC D22S676 Tc2 TC-2 TCII Tcn2 Tcn-2 Tcn2p transcobalamin 2 transcobalamin II transcobalamin II macrocytic anemia transcobalamin-2 vitamin B12-binding protein 2; macrocytic anemia; TC II; TC-2; TCII; Transcobalamin II; transcobalamin II; macrocytic anemia; Transcobalamin-2; vitamin B12-binding protein 2
Gene Aliases: AW208754; D22S676; D22S750; II; TC; TC II; TC-2; TC2; TCII; Tcn-2; TCN2; Tcn2p
UniProt ID: (Human) P20062, (Mouse) O88968
Entrez Gene ID: (Human) 6948, (Mouse) 21452, (Rat) 64365
Molecular Function: cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.