Novus Biologicals
Manufacturer Code:NBP29397820UL
Catalog # NB022522
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 24-280 of human TCN1 (NP_001053.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: haptocorin; haptocorin TC I TC-1 TC1haptocorrin TCItranscobalamin-1 Transcobalamin I transcobalamin I (vitamin B12 binding protein R binder family); Haptocorrin; HC; Protein R; TC I; TC-1; Transcobalamin I; transcobalamin I (vitamin B12 binding protein, R binder family); Transcobalamin-1
Gene Aliases: HC; TC-1; TC1; TCI; TCN1
UniProt ID: (Human) P20061
Entrez Gene ID: (Human) 6947
Molecular Function: cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.