Proteintech
Manufacturer Code:160781AP150UL
Catalog # 50-173-0696
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TCN1 Fusion Protein Ag9072 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: haptocorin; Haptocorrin; HC; Protein R; TC 1 TC I TC1 TCI TCN1 Transcobalamin 1 Transcobalamin I; TC I; TC-1; Transcobalamin I; transcobalamin I (vitamin B12 binding protein, R binder family); Transcobalamin-1
Gene Aliases: HC; TC-1; TC1; TCI; TCN1
UniProt ID: (Human) P20061
Entrez Gene ID: (Human) 6947
Molecular Function: cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.