Invitrogen
Manufacturer Code:PA534419
Catalog # PIPA534419
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 19 amino acid synthetic peptide near the amino terminus of human TCIRG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATPase, H+ transporting, 116kD; ATPase, H+ transporting, lysosomal I; ATPase, H+ transporting, lysosomal V0 protein a; OC-116 kDa; osteoclastic proton pump; Osteoclastic proton pump 116 kDa subunit; osteosclerotic; specific 116-kDa vacuolar proton pump subunit; T cell immune response cDNA7 protein; T-cell immune regulator 1; T-cell immune response cDNA 7; T-cell immune response cDNA7 protein; T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein a; T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein A3; T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 subunit A3; TCIRG1 T-cell immune regulator 1 ATPase H+ transporting lysosomal V0 subunit A3 a3 Stv1 Vph1 Atp6i OC116 OPTB1 TIRC7 ATP6N1C ATP6V0A3 OC-116; TIRC7; V-ATPase 116 kDa subunit a3; V-ATPase 116-kDa; V-ATPase a3; v-H+ATPase subunit a3; V-type proton ATPase 116 kDa subunit a; V-type proton ATPase 116 kDa subunit a isoform 3; V-type proton ATPase 116 kDa subunit a3; vacuolar proton translocating ATPase 116 kDa subunit A; Vacuolar proton translocating ATPase 116 kDa subunit a isoform 3
Gene Aliases: a3; ATP6a3; Atp6i; ATP6N1C; ATP6V0A3; oc; OC-116; OC-116kDa; OC116; OPTB1; Stv1; TCIRG1; TIRC7; Vph1
UniProt ID: (Human) Q13488
Entrez Gene ID: (Human) 10312, (Rat) 293650, (Mouse) 27060
Molecular Function:
ATP synthase
cation transporter
hydrolase
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.