Abnova
Manufacturer Code:H00010312K
Catalog # 89-201-443
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human TCIRG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human TCIRG1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: ATP6N1C ATP6V0A3 Atp6i OC-116kDa OC116 OPTB1 Stv1 TIRC7 Vph1 a3; ATPase, H+ transporting, 116kD; OC-116 kDa; Osteoclastic proton pump 116 kDa subunit; specific 116-kDa vacuolar proton pump subunit; T-cell immune regulator 1; T-cell immune response cDNA 7; T-cell immune response cDNA7 protein; T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein a; T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 subunit A3; TIRC7; V-ATPase 116 kDa subunit a3; V-ATPase 116-kDa; V-type proton ATPase 116 kDa subunit a; V-type proton ATPase 116 kDa subunit a3; vacuolar proton translocating ATPase 116 kDa subunit A; Vacuolar proton translocating ATPase 116 kDa subunit a isoform 3
Gene Aliases: a3; Atp6i; ATP6N1C; ATP6V0A3; OC-116kDa; OC116; OPTB1; Stv1; TCIRG1; TIRC7; Vph1
UniProt ID: (Human) Q13488
Entrez Gene ID: (Human) 10312
Molecular Function: ATP synthase cation transporter hydrolase transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.