Novus Biologicals
Manufacturer Code:NBP119083AF532
Catalog # NP119083A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide made to an internal portion of human TCF7L2 (within residues 150-200). [Swiss-ProtNo. Q9NQB0] |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
Protein Aliases: HMG box transcription factor 4; HMG box transcription factor 4 hTCF-4 T-cell factor 4 T-cell factor-4 variant A T-cell factor-4 variant B T-cell factor-4 variant C T-cell factor-4 variant D T-cell factor-4 variant E T-cell factor-4 variant H T-cell factor-4 variant I T-cell factor-4 variant J T-cell factor-4 variant K T-cell factor-4 variant L T-cell factor-4 variant M T-cell factor-4 variant X2 T-cell-specific transcription factor 4 TCF-4T-cell factor-4 variant F TCF4T-cell factor-4 variant G transcription factor 7-like 2 transcription factor 7-like 2 (T-cell specific HMG-box); hTCF-4; T-cell factor 4; T-cell factor-4 variant A; T-cell factor-4 variant B; T-cell factor-4 variant C; T-cell factor-4 variant D; T-cell factor-4 variant E; T-cell factor-4 variant F; T-cell factor-4 variant G; T-cell factor-4 variant H; T-cell factor-4 variant I; T-cell factor-4 variant J; T-cell factor-4 variant K; T-cell factor-4 variant L; T-cell factor-4 variant M; T-cell factor-4 variant X2; T-cell-specific transcription factor 4; Transcription factor 7-like 2; transcription factor 7-like 2 (T-cell specific, HMG-box)
Gene Aliases: TCF-4; TCF4; TCF7L2
UniProt ID: (Human) Q9NQB0
Entrez Gene ID: (Human) 6934
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.