Invitrogen
Manufacturer Code:PA547810
Catalog # PIPA547810
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit human TCF7L1 Lys429-Ser581 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: HMG box transcription factor 3; TCF-3; TCF3; Transcription factor 7-like 1; transcription factor 7-like 1 (T-cell specific, HMG-box)
Gene Aliases: TCF-3; TCF3; TCF7L1
UniProt ID: (Human) Q9HCS4
Entrez Gene ID: (Human) 83439
Molecular Function: nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.