Invitrogen
Manufacturer Code:PA539499
Catalog # PIPA539499
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human TCF7L1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HMG box transcription factor 3; T-cell factor 3; TCF-3; TCF3; transcription factor 3; Transcription factor 7-like 1; transcription factor 7-like 1 (T-cell specific, HMG box); transcription factor 7-like 1 (T-cell specific, HMG-box); transcription factor 7-like 2 (T-cell specific, HMG box)
Gene Aliases: bHLHb21; TCF-3; TCF3; TCF7L1
UniProt ID: (Human) Q9HCS4, (Mouse) Q9Z1J1
Entrez Gene ID: (Human) 83439, (Mouse) 21415
Molecular Function: nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.