Invitrogen
Manufacturer Code:PA5106757
Catalog # PIPA5106757
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TCF7L1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHb21 HMG box transcription factor 3 T-cell factor 3 Tcf3 TCF-3 Tcf7l1 transcription factor 3 transcription factor 7 like 1 transcription factor 7 like 1 (T cell specific HMG box) transcription factor 7-like 1 transcription factor 7-like 1 (T-cell specific HMG box) transcription factor 7-like 1 (T-cell specific HMG-box) transcription factor 7-like 2 (T-cell specific HMG box); HMG box transcription factor 3; T-cell factor 3; TCF-3; transcription factor 3; Transcription factor 7-like 1; transcription factor 7-like 1 (T-cell specific, HMG box); transcription factor 7-like 1 (T-cell specific, HMG-box); transcription factor 7-like 2 (T-cell specific, HMG box)
Gene Aliases: bHLHb21; TCF-3; TCF3; TCF7L1
UniProt ID: (Human) Q9HCS4, (Mouse) Q9Z1J1
Entrez Gene ID: (Human) 83439, (Rat) 312451, (Mouse) 21415
Molecular Function: nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.