Abnova
Manufacturer Code:H00083439A01
Catalog # 89-107-400
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 489 a.a. &bsim 586 a.a) partial recombinant protein with GST tag.,TCF7L1 (NP_112573 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant TCF7L1.
For Research Use Only
Protein Aliases: HMG box transcription factor 3; TCF-3; TCF3; Transcription factor 7-like 1; transcription factor 7-like 1 (T-cell specific, HMG-box)
Gene Aliases: TCF-3; TCF3; TCF7L1
UniProt ID: (Human) Q9HCS4
Entrez Gene ID: (Human) 83439
Molecular Function: nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.