Invitrogen
Manufacturer Code:MA514965
Catalog # PIMA514965
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | C.725.7 |
Immunogen | Synthetic peptide corresponding to a region surrounding Pro95 of human TCF1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: OTTHUMP00000159390 OTTHUMP00000159391 T-cell-specific transcription factor 1 transcription factor 7 transcription factor-7 T-cell specific; T cell factor-1; T-cell factor 1; T-cell-specific transcription factor 1; TCF-7; Transcription factor 7; transcription factor 7, T-cell specific
Gene Aliases: AI465550; TCF-1; TCF1; TCF7
UniProt ID: (Human) P36402, (Mouse) Q00417
Entrez Gene ID: (Human) 6932, (Mouse) 21414
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.