Invitrogen
Manufacturer Code:PA585281
Catalog # PIPA585281
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human TCF3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHb21; Class B basic helix-loop-helix protein 21; E2A antibody ITF1 antibody MGC129647 antibody MGC129648 antibody VDIR antibody bHLHb21 antibody TCF3 antibody VDR interacting repressor antibody transcription factor ITF-1 antibody negative vitamin D response element-binding protein antibody class B basic helix-loop-helix protein 21 antibody immunoglobulin transcription factor 1 antibody kappa-E2-binding factor antibody helix-loop-helix protein HE47 antibody vitamin D receptor-interacting repressor antibody transcription factor E2-alpha antibody transcription factor 3 (E2A immunoglobulin enhancer binding factors E12/E47) antibody; helix-loop-helix protein HE47; Immunoglobulin enhancer-binding factor E12/E47; Immunoglobulin transcription factor 1; Kappa-E2-binding factor; negative vitamin D response element-binding protein; NOL1-TCF3 fusion; TCF-3; Transcription factor 3; transcription factor 3 (E2A immunoglobulin enhancer binding factors E12/E47); transcription factor 3 variant 3; Transcription factor A1; Transcription factor E2-alpha; transcription factor E2a; Transcription factor ITF-1; VDR interacting repressor; vitamin D receptor-interacting repressor
Gene Aliases: A1; AA408400; AGM8; ALF2; AW209082; BHLHB21; E12; E12/E47; E2A; E47; ITF1; ME2; Pan1; Pan2; TCF-3; TCF3; Tcfe2a; VDIR
UniProt ID: (Human) P15923, (Mouse) P15806
Entrez Gene ID: (Human) 6929, (Mouse) 21423
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.