Invitrogen
Manufacturer Code:PA518641
Catalog # PIPA518641
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (KAPRARTSPDEDED) corresponding to the internal amino acids of TCF3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHb21; Class B basic helix-loop-helix protein 21; helix-loop-helix protein HE47; Immunoglobulin enhancer-binding factor E12/E47; Immunoglobulin transcription factor 1; Kappa-E2-binding factor; negative vitamin D response element-binding protein; NOL1-TCF3 fusion; TCF-3; TCF3 ITF1 transcription factor 3 (E2A immunoglobulin enhancer binding factors E12/E47) E2A MGC129647 MGC129648 E2A immunoglobulin enhancer-binding factor E12/E47 immunoglobulin transcription factor 1 kappa-E2-binding factor transcription factor 3 transcription factor E2-alpha; Transcription factor 3; transcription factor 3 (E2A immunoglobulin enhancer binding factors E12/E47); transcription factor 3 variant 3; Transcription factor E2-alpha; Transcription factor ITF-1; VDR interacting repressor; vitamin D receptor-interacting repressor
Gene Aliases: AGM8; BHLHB21; E2A; E47; ITF1; TCF-3; TCF3; VDIR
UniProt ID: (Human) P15923
Entrez Gene ID: (Human) 6929
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.