Invitrogen
Manufacturer Code:PA572640
Catalog # PIPA572640
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 5-35 amino acids from the N-terminal region of DANRE tcf21 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Capsulin; Epicardin; MyoRa2; TCF-21; TCF21 BHLHA23 POD1 Transcription factor 21 Capsulin Class A basic helix-loop-helix protein 23 Epicardin Podocyte-expressed 1; Transcription factor 21
Gene Aliases: tcf21; zgc:123313
UniProt ID: (Zebrafish) Q32PV5
Entrez Gene ID: (Zebrafish) 558148
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.