Invitrogen
Manufacturer Code:PA568595
Catalog # PIPA568595
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human TCF21 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHa23; Capsulin; Class A basic helix-loop-helix protein 23; Epicardin; Pod-1; Podocyte-expressed 1; TCF-21; TCF21 BHLHA23 POD1 Transcription factor 21 Capsulin Class A basic helix-loop-helix protein 23 Epicardin Podocyte-expressed 1; Transcription factor 21
Gene Aliases: BHLHA23; Capsulin; epc; epicardin; Msf-1; Pod-1; POD1; TCF21
UniProt ID: (Human) O43680, (Bovine) Q5E9S3, (Mouse) O35437
Entrez Gene ID: (Human) 6943, (Horse) 100073150, (Bovine) 510873, (Dog) 483993, (Rabbit) 100346722, (Rat) 252856, (Guinea pig) 100719500, (Mouse) 21412
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.