Novus Biologicals
Manufacturer Code:NBP293154
Catalog # NB028196
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 30-100 of human TCF21 (NP_003197.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHa23; bHLHa23BHLHA23 Capsulin Class A basic helix-loop-helix protein 23 epicardin Pod-1 POD1pod-1 Podocyte-expressed 1 TCF-21 transcription factor 21; Capsulin; Class A basic helix-loop-helix protein 23; Epicardin; Pod-1; Podocyte-expressed 1; TCF-21; Transcription factor 21
Gene Aliases: BHLHA23; POD1; TCF21
UniProt ID: (Human) O43680
Entrez Gene ID: (Human) 6943
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.