Abnova
Manufacturer Code:PAB21931
Catalog # 89-123-639
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human TCF21. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant TCF21.
For Research Use Only
Protein Aliases: bHLHa23; Capsulin; Class A basic helix-loop-helix protein 23; Epicardin; Pod-1; POD1; Podocyte-expressed 1; TCF-21; Transcription factor 21
Gene Aliases: BHLHA23; POD1; TCF21
UniProt ID: (Human) O43680
Entrez Gene ID: (Human) 6943
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.