Invitrogen
Manufacturer Code:PA541482
Catalog # PIPA541482
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human TCF15 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: basic helix-loop-helix transcription factor 15; bHLHa40; Class A basic helix-loop-helix protein 40; Meso1; Paraxis; Protein bHLH-EC2; TCF-15; Transcription factor 15; transcription factor 15 (basic helix-loop-helix); Transcription factor 15 (basic helix-loop-helix) EC2 PARAXIS bHLHa40
Gene Aliases: bHLH-EC2; BHLHA40; BHLHEC2; EC2; Meso1; par1; PARAXIS; TCF15
UniProt ID: (Human) Q12870, (Mouse) Q60756
Entrez Gene ID: (Bovine) 518491, (Dog) 609667, (Horse) 100067919, (Rabbit) 100347808, (Human) 6939, (Mouse) 21407, (Rat) 296272, (Guinea pig) 100715168, (Zebrafish) 30159
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.