Novus Biologicals
Manufacturer Code:NBP2836300.1ML
Catalog # NB031559
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of human TCF15 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: basic helix-loop-helix transcription factor 15; bHLHa40; Class A basic helix-loop-helix protein 40; Paraxis; Protein bHLH-EC2; TCF-15; Transcription factor 15; transcription factor 15 bHLHa40 EC2 PARAXIS transcription factor 15 (basic helix-loop-helix)
Gene Aliases: BHLHA40; BHLHEC2; EC2; PARAXIS; TCF15
UniProt ID: (Human) Q12870
Entrez Gene ID: (Human) 6939
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.