Invitrogen
Manufacturer Code:PA5102748
Catalog # PIPA5102748
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TCEB2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 0610040H15Rik 18 kDa elongin B eloB Elongin 18 kDa subunit elongin B elongin 18-kD subunit elongin-B RNA polymerase II transcription factor SIII p18 subunit RNA polymerase II transcription factor SIII subunit B SIII SIII p18 TCEB 2 TCEB2 transcription elongation factor B (SIII) polypeptide 2 (18kD elongin B) transcription elongation factor B (SIII) polypeptide 2 transcription elongation factor B (SIII) polypeptide 2 (18 kDa elongin B) transcription elongation factor B (SIII) polypeptide 2 (18kDa elongin B) transcription elongation factor B polypeptide 2 transcription elongation factor B subunit 2; 18 kDa, elongin B; EloB; Elongin 18 kDa subunit; elongin B; elongin, 18-kD subunit; Elongin-B; RNA polymerase II transcription factor SIII p18 subunit; RNA polymerase II transcription factor SIII subunit B; SIII p18; transcription elongation factor B (SIII) polypeptide 2 (18kD, elongin B); transcription elongation factor B (SIII), polypeptide 2 (18 kDa, elongin B); transcription elongation factor B (SIII), polypeptide 2 (18kDa, elongin B); Transcription elongation factor B polypeptide 2
Gene Aliases: 0610040H15Rik; ELOB; SIII; TCEB2
UniProt ID: (Human) Q15370, (Mouse) P62869, (Rat) P62870
Entrez Gene ID: (Human) 6923, (Mouse) 67673, (Rat) 81807
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.